Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: entactin 2; Entactin-2; NID-2; nidogen 2 (osteonidogen); Nidogen-2; Osteonidogen
Gene Aliases: AW547149; entactin-2; Ly111; NID-2; NID2; nidogen-2
UniProt ID: (Human) Q14112, (Mouse) O88322, (Rat) B5DFC9
Entrez Gene ID: (Human) 22795, (Mouse) 18074, (Rat) 302248
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support