Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitocondrial matrix and uses thiamine pyrophosphate as a cofactor. A congential deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoglutarate dehydrogenase complex component E1; 2-oxoglutarate dehydrogenase E1 component, mitochondrial; 2-oxoglutarate dehydrogenase, mitochondrial; Alpha-ketoglutarate dehydrogenase; Alpha-KGDH-E1; E1o; OGDC-E1; oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide); oxoglutarate decarboxylase; oxoglutarate dehydrogenase (lipoamide); oxoglutarate dehydrogenase (succinyl-transferring); testicular tissue protein Li 131; Thiamine diphosphate (ThDP)-dependent 2-oxoglutarate dehydrogenase
Gene Aliases: AKGDH; E1k; OGDC; OGDH; Ogdhl
UniProt ID: (Human) Q02218, (Rat) Q5XI78
Entrez Gene ID: (Human) 4967, (Rat) 360975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support