Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCQALQCQD LPVPNEARVN CSHPFGAFRY QSVCSFTCNE GLLLVGASVL QCLATGNWNS VPPECQAIPC TPLLSPQNGT MTCVQPLGSS SYKSTCQFIC DEGYSLSGPE RLDCTRSGRW TDSPPMCEAI KCPELFAPEQ GSLDCSDTRG EFNVGSTCHF SCNNGFKLEG PNNVECTTSG RWSATPPTCK GIASLPTPGV QCPALTTPGQ GTMYCRHHPG TFGFNTTCYF GCNAGFTLIG DSTLSCRPSG QWTAVTPACR AVKCSELHVN KPIAMNCSNL WGNFSYGSIC SFHCLEGQLL NGSAQTACQE NGHWSTTVPT CQDDGKCPLN PHSHLGTYGV FTNAAFDPSP (481-830 aa encoded by BC028067)
P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate. P-selectin protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. P-selectin plays an important role in adhesive processes between leucocytes and endothelial cells, and is a calcium dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens found on neutrophils and monocytes. P-selectin is constitutively expressed in inflammation and contributes to atherogenesis, thrombosis and tissue destruction. Clinically, P-selectin is used to distinguish heparin induced thrombocytopenia with and without thrombosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD62 antigen-like family member P; CD62P; FLJ45155; GMP-140; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; LECAM3; Leukocyte-endothelial cell adhesion molecule 3; P-selectin; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; RP1-86F14.2; selectin P (granule membrane protein 140kDa, antigen CD62)
Gene Aliases: CD62; CD62P; GMP140; GMRP; GRMP; LECAM3; PADGEM; PSEL; SELP
UniProt ID: (Human) P16109
Entrez Gene ID: (Human) 6403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support