Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: Mouse brain tissue lysates, rat brain tissue, human esophagus tissue, mouse brain tissue, MCF-7.
Subcellular Location: Cell membrane.
P2rx2 are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. P2rx2 functions as a ligand-gated ion channel and binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for P2rx2. P2rx2 is involved in a variety of cellular responses, such as excitatory postsynaptic responses in sensory neurons, neuromuscular junctions (NMJ) formation, hearing, perception of taste and peristalsis. In the inner ear, P2rx2 regulates sound transduction and auditory neurotransmission, outer hair cell electromotility, inner ear gap junctions, and K(+) recycling. P2rx2 also mediates synaptic transmission between neurons and from neurons to smooth muscle. Diseases associated with P2RX2 include Deafness, Autosomal Dominant 41 and Dfna41 Nonsyndromic Hearing Loss And Deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP receptor; MGC129601; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2
Gene Aliases: DFNA41; P2RX2; P2X2; P2X2a
UniProt ID: (Human) Q9UBL9, (Rat) P49653, (Mouse) Q8K3P1
Entrez Gene ID: (Human) 22953, (Rat) 114115, (Mouse) 231602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support