Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Group IB phospholipase A2; MGC119834; MGC119835; Phosphatidylcholine 2-acylhydrolase 1B; Phospholipase A2; PLA2-Ib
Gene Aliases: Pla2; Pla2a; Pla2g1b; sPLA2IB
UniProt ID: (Mouse) Q9Z0Y2
Entrez Gene ID: (Mouse) 18778
Molecular Function: phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support