Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLLYAAEIL EIEYLEEQCL KMLETIQASD DNDTEATMAD GGAEEEEDRK ARYLKNIFIS KHSSEESGYA SVAGQSLPGP MVDQSPSVST SFGLSAMSPT KAAVDSLMTI GQSLLQGTLQ PPAGPEEPTL AGGGRHPGVA EVKTEMMQVD EVPSQDSPGA AESSISGGMG DKVEERGKEG PGTPTRSSVI TSARELHYGR EESAEQVPPP AEAGQAPTGR PEHPAPPPEK HLGIYSVLPN HKADAVLSMP SSVTSGLHVQ PALAVSMDFS TYGGLLPQGF IQRELFSKLG ELAVGMKSES R (101-400 aa encoded by BC026902)
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; zinc finger and BTB domain containing 16 protein; Zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); Zinc finger protein PLZF
Gene Aliases: Lx; PLZF; ZBTB16; Zfp145; ZNF145
UniProt ID: (Human) Q05516
Entrez Gene ID: (Human) 7704, (Rat) 353227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support