Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Antibody detects endogenous levels of CSK only when phosphorylated at Tyr304.
CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene. CSK plays an important role in the regulation of cell growth, differentiation, migration, and immune responses. CSK phosphorylates tyrosine residues located in the C-terminal tails of Src-family kinases including LCK, SRC, HCK, FYN, LYN, CSK, and YES1. CSK is expressed ubiquitously as a protein of approximately 50 KDa in cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-SRC kinase; MGC117393; p50CSK; protein-tyrosine kinase (CSK); Protein-tyrosine kinase CYL; Protein-tyrosine kinase MPK-2; Tyrosine-protein kinase CSK
Gene Aliases: AW212630; CSK; p50CSK
UniProt ID: (Human) P41240, (Rat) P32577, (Mouse) P41241
Entrez Gene ID: (Human) 1445, (Rat) 315707, (Mouse) 12988
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support