Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Antibody detects endogenous levels of DYRK1A/B only when phosphorylated at Tyr321/273.
DYRK1A is a member of the dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) family and is thought to be involved in neuronal differentiation, neurodegenerative diseases, and is considered to be a strong candidate gene for learning defects associated with Down syndrome. DYRK1A phosphorylates several transcription factors such as p53, leading to inhibition of embryonic neuronal cell proliferation. DYRK1A associates with multiple proteins, including SPRED1 and SPRED2; this association appears to inhibit the phosphorylation activity of DYRK1A. Recent evidence also suggests that DYRK1A may be involved in the replication of HIV-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; Dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual specificity tyrosine-phosphorylation-regulated kinase 1B; Dual Specificity Yak1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; hMNB; HP86; Minibrain-related kinase; Mirk protein kinase; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; Protein kinase minibrain homolog; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Aliases: 2310043O08Rik; AOMS3; D16Ertd272e; D16Ertd493e; DYRK; DYRK1; DYRK1A; DYRK1B; ENSMUSG00000074897; Gm10783; HP86; MIRK; mmb; MNB; MNBH; Mp86; MRD7; PSK47
UniProt ID: (Human) Q13627, (Human) Q9Y463, (Mouse) Q61214, (Rat) Q63470, (Mouse) Q9Z188
Entrez Gene ID: (Human) 1859, (Human) 9149, (Mouse) 13548, (Rat) 25255, (Mouse) 13549, (Rat) 308468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support