Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Antibody detects endogenous levels of DYRK2/4 only when phosphorylated at Y382/Y264.
DYRK2 is a member of the dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) family that is thought to be involved in cellular proliferation and apoptosis. Under normal conditions, nuclear but not cytoplasmic DYRK2 is ubiquitinated by MDM2, leading to its constitutive degradation. However, upon exposure to genotoxic stress, ATM phosphorylates DYRK2, leading to its dissociation from MDM2 and its phosphorylation of p53, thereby inducing apoptosis. Recent evidence also suggests that DYRK2 may serve as a scaffold that facilitates assembly of an E3 ubiquitin ligase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; Dual specificity tyrosine-phosphorylation-regulated kinase 2; Dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dyrk2; dyrk4; RAD51 associated protein 1
Gene Aliases: 1810038L18Rik; AW049118; DYRK2; DYRK4; Dyrk4a; Dyrk4b; Rad51ap1
UniProt ID: (Human) Q92630, (Human) Q9NR20, (Mouse) Q5U4C9, (Mouse) Q8BI55
Entrez Gene ID: (Human) 8445, (Human) 8798, (Mouse) 69181, (Rat) 314862, (Mouse) 101320, (Rat) 312721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support