Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAPK7 (ERK5) serine/threonine kinase is a member of the MAP kinase family and is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. MAPK7 is activated by mitogen-activated protein kinase 5 (MAP2K5/MEK5) and is involved in downstream signaling processes of various receptors. In response to extracelluar signals, MAPK7 translocates to the cell nucleus where it regulates gene expression by phosphorylating and activating different transcription factors. Gene deletion of MAPK7 in mice results in defective blood vessel and cardiac development leading to embryonic lethality. MAPK7 has been shown to be critical for endothelial function and maintenance of blood vessel integrity. Four alternatively spliced transcript variants of this gene encoding two distinct isoforms have been reported. ERK5 is expressed in many adult tissues, abundantly in heart, placenta, lung, kidney and skeletal muscle, but is not detectable in liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Big MAP kinase 1; BMK-1; BMK1 kinase; BNhp-1; BNhp1 kinase; ERK-5; extracellular signal regulated kinase 5; Extracellular signal-regulated kinase 5; extracellular-signal-regulated kinase 5; MAP kinase 7; MAPK 7; mitogen activated protein kinase 7; Mitogen-activated protein kinase 7
Gene Aliases: BMK-1; BMK1; ERK-5; ERK4; ERK5; Erk5-T; MAPK7; PRKM7
UniProt ID: (Human) Q13164, (Mouse) Q9WVS8
Entrez Gene ID: (Human) 5598, (Mouse) 23939, (Rat) 114509
Molecular Function:
non-receptor serine/threonine protein kinase
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support