Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-GAB2 (Tyr614) Polyclonal Antibody detects endogenous levels of GAB2 only when phosphorylated at Tyr614.
This gene is a member of the GRB2-associated binding protein (GAB) gene family. These proteins contain pleckstrin homology (PH) domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Grb2 associated binder 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; PH domain-containing adaptor molecule p97; pp100
Gene Aliases: AI463667; D130058I17Rik; GAB2; KIAA0571; p97
UniProt ID: (Human) Q9UQC2, (Rat) Q9EQH1
Entrez Gene ID: (Human) 9846, (Rat) 84477, (Mouse) 14389
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support