Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with human and rat based on sequence homology.
ICK or intestinal cell kinase belongs to a very extensive family of proteins that share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. ICK encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. ICK localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. ICK purified from transfected human embryonic kidney cells undergoes autophosphorylation and phosphorylates a test protein. ICK plays a key role in development of central nervous, skeletal, and endocrine systems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliogenesis associated kinase 1; heart serine-threonine protein kinase; hICK; Intestinal cell kinase; Laryngeal cancer kinase 2; LCK2; MAK-related kinase; mICK; MRK; serine/threonine protein kinase; Serine/threonine-protein kinase ICK
Gene Aliases: 2210420N10Rik; AI848300; CILK1; ECO; ICK; KIAA0936; LCK2; MRK
UniProt ID: (Human) Q9UPZ9, (Rat) Q62726, (Mouse) Q9JKV2
Entrez Gene ID: (Human) 22858, (Rat) 84411, (Mouse) 56542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support