Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of JAK2/3 only when phosphorylated at Tyr966/939.
Janus Activating Kinase 2 and 3 (JAK2/JAK3) is one of a number of related tyrosine kinases involved in cytoplasmic signal transduction. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Breast-JAK3; JAK-2; JAK-3; JAK3B; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L-JAK; Leukocyte janus kinase; Tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3
Gene Aliases: fae; Fd17; JAK-3; JAK2; JAK3; JAK3_HUMAN; JAKL; JTK10; L-JAK; LJAK; RATJAK3; THCYT3; wil
UniProt ID: (Human) O60674, (Human) P52333, (Rat) Q62689, (Mouse) Q62120, (Mouse) Q62137, (Rat) Q63272
Entrez Gene ID: (Human) 3717, (Human) 3718, (Rat) 24514, (Mouse) 16452, (Mouse) 16453, (Rat) 25326
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support