Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody was cross-adsorbed to phospho-WAVE (Tyr-150) and unphosphorylated WAVE (Tyr-125) peptides before affinity purification using phospho-WAVE (Tyr-125) peptide. In western blots, the antibody detects an 80 kDa* band corresponding to phosphorylated WAVE in human SYF cSrc transformed cells and mouse macrophages treated with pervanadate, but is not observed in control cells. These bands can be specifically blocked with phospho-WAVE (Tyr-125) peptide (WX1775), but not phospho-WAVE (Tyr-150) peptide (WX1825).
WASP (for Wiskott-Aldrich syndrome protein) and N-WASP are downstream effectors of Cdc42 that are implicated in actin polymerization and cytoskeletal organization. The WASP family also includes VASP (vasodilator-stimulated phosphoprotein) and Mena (for mammalian enabled protein), which accumulate at focal adhesions and are also involved in the regulation of the actin cytoskeleton. The WAVE proteins are related to the WASP family proteins and are likewise involved in mediating actin reorganization downstream of the Rho family of small GTPases. The two protein homologs WAVE1 and WAVE2 specifically regulate membrane ruffling by inducing the formation of actin filament clusters in response to GTP binding and activating Rac. The WAVE proteins mediate this actin polymerization by cooperating with the Arp2/3 complex, a nucleation core, and thereby promoting the formation of actin filaments. WAVE1, which is also designated SCAR (for suppressor of cAR), is expressed primarily in the brain, while WAVE2 is widely expressed with the expression highest in peripheral blood leukocytes. WAVE3 forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-binding protein WASF1; homology of dictyostelium scar 1; Protein WAVE-1; Verprolin homology domain-containing protein 1; WAS protein family; WAS protein family, member 1; WASP family 1; WASP family protein member 1; Wiskott-Aldrich syndrome protein family member 1
Gene Aliases: AI195380; AI838537; KIAA0269; Scar; SCAR1; WASF1; WAVE; WAVE-1; WAVE1
UniProt ID: (Human) Q92558, (Rat) Q5BJU7, (Mouse) Q8R5H6
Entrez Gene ID: (Human) 8936, (Rat) 294568, (Mouse) 83767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support