Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP4 is one of many proteins that interact with and regulate Rab GTPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arfophilin-2; FIP4-Rab11; mRab11-FIP4A; RAB11 family interacting protein 4 (class II); Rab11 family-interacting protein 4
Gene Aliases: A730072L08Rik; ARFO2; KIAA1821; mKIAA1821; RAB11-FIP4; RAB11FIP4; RGD1305063
UniProt ID: (Human) Q86YS3, (Mouse) Q8BQP8
Entrez Gene ID: (Human) 84440, (Mouse) 268451, (Rat) 303337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support