Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: DCPHTIGVEF GTRIIEVSGQ KIKLQIWDTA GQERFRAVTR SYYRGAAGAL MVYDITRRST YNHLSSWLTD ARNLTNPNTV IILIGNKADL EAQRDVTYEE AK
Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA165P4.3 (member RAS oncogene family); F protein-binding protein 1; Ras-related protein Rab-14; small GTP binding protein RAB14
Gene Aliases: 0610030G24Rik; 2810475J17Rik; A830021G03Rik; AI314285; AI649155; D030017L14Rik; FBP; RAB-14; RAB14
UniProt ID: (Human) P61106, (Mouse) Q91V41
Entrez Gene ID: (Human) 51552, (Mouse) 68365
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support