Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide shares 95% identity with RAB1B and is homologous in rat and mouse.
Specificity of this antibody: RAB1.
This gene encodes a member of the Ras superfamily of GTPases. Members of the gene family cycle between inactive GDP-bound and active GTP-bound forms. This small GTPase controls vesicle traffic from the endoplasmic reticulum to the Golgi apparatus. Multiple alternatively spliced transcript variants have been identified for this gene which encode different protein isoforms. [provided by RefSeq, Oct 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP binding protein Rab1a; Rab GTPase YPT1 homolog; RAB1, member RAS oncogene family; RAB1B-like, member RAS oncogene family; Ras-related protein Rab-1A; Ras-related protein Rab-1B; ras-related YPT1 protein; small GTP-binding protein; YPT1-related protein
Gene Aliases: 1110011F09Rik; Ac2-048; Gtbp; mKIAA3012; Rab-1; RAB1; RAB1A; RAB1B; Rab1bl; Rab1r; YPT1
UniProt ID: (Human) P62820, (Human) Q9H0U4, (Mouse) P62821, (Rat) Q6NYB7, (Mouse) Q9D1G1
Entrez Gene ID: (Human) 5861, (Human) 81876, (Mouse) 19324, (Rat) 81754, (Rat) 100126191, (Mouse) 76308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support