Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MTDGDYDYLI KLLALGDSGV GKTTFLYRYT DNKFNPKFIT TVGIDFREKR VVYNAQGPNG SSGKAFKVHL QLWDTAGQER FRSLTTAFFR DAMGFLLMFD LTSQQSFLNV RSWMSQLQAN AYCENPDIVL IGNKADLPDQ REVNERQARE LADKYGIPYF ETSAATGQNV EKAVETLLDL IMKRMEQCVE KTQIPDTVNG GNSGNLDGEK PPEKKCIC
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C25KG; Ras-related protein Rab-27B
Gene Aliases: C25KG; RAB27B
UniProt ID: (Human) O00194
Entrez Gene ID: (Human) 5874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support