Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP binding protein; GTP-binding protein; OTTMUSP00000004796; rab 2; RAB2, member RAS oncogene family; Ras-related protein Rab-2A; RP23-183L6.1; small GTP binding protein RAB2A
Gene Aliases: 9330148M11Rik; C80220; LHX; RAB2; RAB2A
UniProt ID: (Human) P61019, (Mouse) P53994, (Rat) P05712
Entrez Gene ID: (Human) 5862, (Mouse) 59021, (Rat) 65158
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support