Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma-aminobutyric acid receptor subunit rho1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; OPN 2; opsin 2; opsin 2, rod pigment; Opsin-2; Red Opsin; Rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); Rod Opsin; rod visual pigment; RP 4
Gene Aliases: CSNBAD1; Noerg1; OPN2; Ops; PIGRHO1; RHO; RHO1; RP4
UniProt ID: (Bovine) P02699, (Pig) O18766, (Human) P08100, (Rat) P51489, (Mouse) P15409
Entrez Gene ID: (Bovine) 509933, (Pig) 397437, (Human) 6010, (Horse) 100056641, (Rat) 24717, (Mouse) 212541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support