Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with Monkey, Pig, Cat, Mouse.
SOS2 promotes the exchange of Ras-bound GDP by GTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Son of sevenless homolog 2; SOS-2
Gene Aliases: NS9; SOS-2; SOS2
UniProt ID: (Human) Q07890
Entrez Gene ID: (Human) 6655
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support