Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGPRTSSTS SSVNSQTLNR NQPTVYIAAS PPNTDELMSR SQPKVYISAN AATGDEQVMR NQPTLFISTN SGASAASRNM SGQVSMGPAF IHHHPPKSRA IGNNSATSPR VVVTQPNTKY TFKITVSPNK PPAVSPGVVS PTFELTNLLN HPDHYVETEN IQHLTDPTLA HVDRISETRK LSMGSDDAAY TQALLVHQKA RMERLQRELE IQKKKLDKLK SEVNEMENNL TRRRLKRSNS ISQIPSLEEM QQLRSCNRQL QIDIDCLTKE IDLFQARGPH FNPSAIHNFY DNIGFVGPVP PKPKDQRSII KTPKTQDTED DEGAQWNCTA CTFLNHPALI RCEQCEMPRH F (344-693 aa encoded by BC035910)
TAB2 is an activator of MAP3K7/TAK1, which is required for the IL-1 induced activation NF-kappaB and MAPK8/JNK. This protein forms a kinase complex with TRAF6, MAP3K7 and TAB1, thus serves as an adaptor linking MAP3K7 and TRAF6. This protein, TAB1, and MAP3K7 also participate in the signal transduction induced by TNFSF11/RANKL through the activation of the receptor activator of NF-kappaB (TNFRSF11A/RANK), which may regulate the development and function of osteoclasts. Recent experiments have shown that TAB2 and the related protein TAB3 constitutively interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21885; mitogen activated protein kinase kinase kinase 7 interacting protein 2; mitogen-activated protein kinase kinase kinase 7 interacting protein 2; Mitogen-activated protein kinase kinase kinase 7-interacting protein 2; OTTHUMP00000017388; TAB-2; Tak1 binding protein 2; TAK1-binding protein 2; TGF-beta-activated kinase 1 and MAP3K7-binding protein 2; TGF-beta-activated kinase 1-binding protein 2
Gene Aliases: 1110030N06Rik; A530078N03Rik; CHTD2; KIAA0733; MAP3K7IP2; mKIAA0733; TAB-2; TAB2
UniProt ID: (Human) Q9NYJ8, (Mouse) Q99K90
Entrez Gene ID: (Human) 23118, (Mouse) 68652, (Rat) 308267
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support