Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLRVVEGIFI FVVVSESVFG VLGNGFIGLV NCIDCAKNKL STIGFILTGL AISRIFLIWI IITDGFIQIF SPNIYASGNL IEYISYFWVI GNQSSMWFAT SLSIFYFLKI ANFSNYIFLW LKSG
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC126811; MGC126813; T2R10; Taste receptor family B member 2; Taste receptor type 2 member 10; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2
Gene Aliases: T2R10; TAS2R10; TRB2
UniProt ID: (Human) Q9NYW0
Entrez Gene ID: (Human) 50839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support