Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQSTSNHLWL LSDILGQGAT ANVFRGRHKK TGDLFAIKVF NNISFLRPVD VQMREFEVLK KLNHKNIVKL FAIEEETTTR HKVLIMEFCP CGSLYTVLEE PSNAYGLPES EFLIVLRDVV GGMNHLRENG IVHRDIKPGN IMRVIGEDGQ SVYKLTDFGA ARELEDDEQF VSLYGTEEYL HPDMYERAVL RKDHQKKYGA TVDLWSIGVT FYHAATGSLP FRPFEGPRRN KEVMYKIITG KPSGAISGVQ KAENGPIDWS GDMPVSCSLS RGLQVLLTPV LANILEADQE KCWGFDQFFA ETSDILHRMV IHVFSLQQMT AHKIYIHSYN TATIFHELVY KQTKIISSNQ ELIYEGRRLV LEPGRLAQHF PKTTEENPIF VVSREPLNTI GLIYEKISLP KVHPRYDLDG DASMAKAITG VVCYACRIAS TLLLYQELMR KGIRWLIELI KDDYNETVHK KTEVVITLDF CIRNIEKTVK VYEKLMKINL EAAELGEISD IHTKLLRLSS SQGTIETSLQ DIDSRLSPGG SLADAWAHQE GTHPKDRNVE KLQVLLNCMT EIYYQFKKDK AERRLAYNEE QIHKFDKQKL YYHATKAMTH FTDECVKKYE AFLNKSEEWI RKMLHLRKQL LSLTNQCFDI EEEVSKYQEY TNELQETLPQ KMFTASSGIK HTMTPIYPSS NTLVEMTLGM KKLKEEMEGV VKELAENNHI LERFGSLTMD GGLRNVDCL
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling. TBK1 is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-KB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-KB is associated with IKB proteins in the cell cytoplasm, which inhibit NF-KB activity. Phosphorylation of I-KB by IKB kinase (IKK) complex leads to degradation of I-KB and activation of NF-KB. The IKK complex contains IKKa, IKK beta, and IKKK. A novel IKK related kinase was recently identified and designated TBK1 (TANK-binding kinase 1), NAK (NF-KB-activating kinase), and T2K. NAK/TBK1 activates IKK beta through direct phosphorylation. NAK/TBK1 is activated by growth factors and PMA and mediates IKK and NF-kB activation in response to growth factors. NAK/TBK1 functions upstream of NIK and the IKK complex. NAK/TBK1 is also critical in protecting embryonic liver from apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11330; NF-kappa-B-activating kinase; NF-kB-activating kinase; Serine/threonine-protein kinase TBK1; T2K; TANK-binding kinase 1; tbk
Gene Aliases: FTDALS4; NAK; T2K; TBK1
UniProt ID: (Human) Q9UHD2
Entrez Gene ID: (Human) 29110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support