Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MADADEGFG LAHTPLEPDA KDLPCDSKPE SALGAPSKSP SSPQAAFTQQ GMEGIKVFLH ERELWLKFHE VGTEMIITKA GRRMFPSYKV KVTGLNPKTK YILLMDIVPA DDHRYKFADN KWSVTGKAEP AMPGRLYVHP DSPATGAHWM RQLVSFQKLK LTNNHLDPFG HIILNSMHKY QPRLHIVKAD ENNGFGSKNT AFCTHVFPET AFIAVTSYQN HKITQLKIEN NPFAKGFRGS DDMELHRMSR MQSKEYPVVP RSTVRQKVAS NHSPFSSESR ALSTSSNLGS QYQCENGVSG PSQDLLPPPN PYPLPQEHSQ IYHCTKRKE (1-328 aa encoded by BC027942 )
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is closely linked to related family member T-box 3 (ulnar mammary syndrome) on human chromosome 12. The encoded protein may play a role in heart development and specification of limb identity. Mutations in this gene have been associated with Holt-Oram syndrome, a developmental disorder affecting the heart and upper limbs. Several transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box protein 5; T-box transcription factor TBX5
Gene Aliases: HOS; TBX5
UniProt ID: (Human) Q99593, (Mouse) P70326, (Rat) Q5I2P1
Entrez Gene ID: (Human) 6910, (Mouse) 21388, (Rat) 304514
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support