Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brachyury protein; Protein T; RP11-459F1.1; T brachyury homolog; T, brachyury homolog; T-box transcription factor T
Gene Aliases: SAVA; T; TBXT; TFT
UniProt ID: (Human) O15178
Entrez Gene ID: (Human) 6862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support