Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERVGCTLTT TYAHPRPTPT NFLPAISTMA SSYRDRFPHS NLTHSLSLPW RPSTYYKVAS NSPSVAPYCT RSQRVSENTM LPFVSNRTTF FTRYTPDDWY RSNLTNYQES NTSRHNSEKL RVDTSRLIQD KYQQTRKTQA DTTQNLGERV NDIGFWKSEI IHELDEMIGE TNALTDVKKR LERALMETEA PLQVARECLF HREKRMGIDL VHDEVEAQLL TEVDTILCCQ ERMKLHLDKA IAQLAANRAS QHELEKDLSD KQTAYRINDK CHHLRNTSDG VGYFRGVERV DATVSVPESW AKFTDDNILR SQSERAASAK LRDDIENLLV VTANEMWNQF NKVNLSFTNR IAETADAKNK IQTHLAKTLQ EIFQTEMTIE SIKKAIKDKT AFLKVAQTRL DERTRRPNIE LCRDMAQLRL VNEVHEVDDT IQTLQQRLRD AEDTLQSLVH IKATLEYDLA VKANSLYIDQ EKCMSMRKSY PNTLRLVGFC
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tektin-3; testicular microtubules-related protein
Gene Aliases: TEKT3
UniProt ID: (Human) Q9BXF9
Entrez Gene ID: (Human) 64518
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support