Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGLAWGLGVL FLMHVCGTNR IPESGGDNSV FDIFELTGAA RKGSGRRLVK GPDPSSPAFR IEDANLIPPV PDDKFQDLVD AVRAEKGFLL LASLRQMKKT RGTLLALERK DHSGQVFSVV SNGKAGTLDL SLTVQGKQHV VSVEEALLAT GQWKSITLFV QEDRAQLYID CEKMENAELD VPIQSVFTRD LASIARLRIA KGGVNDNFQG VLQNVRFVFG TTPEDILRNK GCSSSTSVLL TLDNNVVNGS SPAIRTNYIG HKTKDLQAIC GISCDELSSM VLELRGLRTI VTTLQDSIRK VTEENKELAN ELRRPPLCYH NGVQYRNNEE WTVDSCTECH CQNSVTICKK VSCPIMPCSN ATVPDGECCP RCWPSDSADD GWSPWSEWTS CSTSCGNGIQ QRGRSCDSLN NRCEGSSVQT RTCHIQECDK RFKQDGGWSH WSPWSSCSVT CGDGVITRIR LCNSPSPQMN GKPCEGEARE TKACKKDACP INGGWGPWSP WDICSVTCGG GVQKRSRLCN NPTPQFGGKD CVGDVTENQI CNKQDCPIDG CLSNPCFAGV KCTSYPDGSW KCGACPPGYS GNGIQCTDVD ECKEVPDACF NHNGEHRCEN TDPGYNCLPC PPRFTGSQPF GQGVEHATAN KQVCKPRNPC TDGTHDCNKN AKCNYLGHYS DPMYRCECKP GYAGNGIICG EDTDLDGWPN ENLVCVANAT YHCKKDNCPN LPNSGQEDYD KDGIGDACDD DDDNDKIPDD RDNCPFHYNP AQYDYDRDDV GDRCDNCPYN HNPDQADTDN NGEGDACAAD IDGDGILNER DNCQYVYNVD QRDTDMDGVG DQCDNCPLEH NPDQLDSDSD RIGDTCDNNQ DIDEDGHQNN LDNCPYVPNA NQADHDKDGK GDACDHDDDN DGIPDDKDNC RLVPNPDQKD SDGDGRGDAC KDDFDHDSVP DIDDICPENV DISETDFRRF QMIPLDPKGT SQNDPNWVVR HQGKELVQTV NCDPGLAVGY DEFNAVDFSG TFFINTERDD DYAGFVFGYQ SSSRFYVVMW KQVTQSYWDT NPTRAQGYSG LSVKVVNSTT GPGEHLRNAL WHTGNTPGQV RTLWHDPRHI GWKDFTAYRW RLSHRPKTGF IRVVMYEGKK IMADSGPIYD KTYAGGRLGL FVFSQEMVFF SDLKYECRDP
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycoprotein G; Thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1
Gene Aliases: THBS; THBS-1; THBS1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996
Entrez Gene ID: (Human) 7057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support