Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 2
TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTU domain-containing protein 7C; Putative DNA-binding protein A20; RP11-356I2.3; TNF alpha-induced protein 3; TNFAIP3 (A20); Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; Zinc finger protein A20
Gene Aliases: A20; AISBL; OTUD7C; TNFA1P2; TNFAIP3
UniProt ID: (Human) P21580
Entrez Gene ID: (Human) 7128, (Rat) 683206
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support