Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KMDSPGALQ TNPPLKLHTD RSAGTPVFVP EQGGYKEKFV KTVEDKYKCE KCHLVLCSPK QTECGHRFCE SCMAALLSSS SPKCTACQES IVKDKVFKDN CCKREILALQ IYCRNESRGC AEQLMLGHLL VHLKNDCHFE ELPCVRPDCK EKVLRKDLRD HVEKACKYRE ATCSHCKSQV PMIALQKHED TDCPCVVVSC PHKCSVQTLL RSELSAHLSE CVNAPSTCSF KRYGCVFQGT NQQIKAHEAS SAVQHVNLLK EWSNSLEKKV SLLQNESVEK NKSIQSLHNQ ICSFEIEIER QKEMLRNNES KILHLQRVID SQAEKLKELD KEIRPFRQNW EEADSMK (6-351 aa encoded by BC075087)
Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e. g. RANK, CD30, CD40, etc. ) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6). TRAF3, originally named CRAF1, interacts directly with the CD40 cytoplasmic tail through a region of similarity to the tumor necrosis factor-alpha (TNF-alpha) receptor-associated factors. TRAF3 binds only a single site, which contains the sequence PEQET, whereas TRAF1 and TRAF2 are capable of binding to either the PEQET site or an additional downstream domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD40 associated protein 1; CD40 binding protein; CD40 receptor associated factor 1; CD40 receptor-associated factor 1; CD40-binding protein; CD40BP; CRAF1; LAP1; LMP1-associated protein 1; RING-type E3 ubiquitin transferase TRAF3; TNF receptor-associated factor 3; TRAFAMN
Gene Aliases: AI528849; amn; CAP-1; CAP1; CD40bp; CRAF1; IIAE5; LAP1; T-BAM; TRAF3; TRAFAMN
UniProt ID: (Human) Q13114, (Mouse) Q60803
Entrez Gene ID: (Human) 7187, (Mouse) 22031
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support