Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Aliquoting is unnecessary for -20°C storage.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycoprotein G; Thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1
Gene Aliases: tbsp1; THBS; THBS-1; THBS1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996
Entrez Gene ID: (Human) 7057, (Rat) 445442, (Mouse) 21825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support