Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QGEVIDIRGP IRDTLQNCCP LPARIQEIVV ETPTLAAERE RSQESPNTPA PPLSMLRIKS ENGEQAFLLM MQPDNTIGDV RALLAQARVM DASAFEIFS
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colorectal tumor-associated antigen COA-1; colorectal tumor-associated antigen-1; Socius; UBX domain-containing protein 11; UBX domain-containing protein 5
Gene Aliases: COA-1; PP2243; SOC; SOCI; UBXD5; UBXN11
UniProt ID: (Human) Q63HM6
Entrez Gene ID: (Human) 91544
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support