Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MAAPPARADA DPSPTSPPTA RDTPGRQAEK SETACEDRSN AESLDRLLPP VGTGRSPRKR TTSQCKSEPP LLRTSKRTIY TAGRPPWYNE HGTQSKEAFA IGLGGGSASG KTTVARMIIE ALDVPWVVLL SMDSFYKVLT EQQQEQAAHN NFNFDHPDAF DFDLIISTLK KLKQGKSVKV PIYDFTTHSR KKDWKTLYGA NVIIFEGIMA FADKTLLELL DMKIFVDTDS DIRLVRRLRR DISERGRDIE GVIKQYNKFV KPSFDQYIQP TMRLADIVVP RGSGNTVAID LIVQHVHSQL EEGCAGLGTP VPPAAPDAER PEEHAAGTGH AHHHQVRAHL GTGRPRARAP AQLLAPQGQG DQSRRVHLLL QETDAAAHRA RALLPALSGL RRTDPAGAGL CGQVLCGEAD HRCVHSARR
Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UCK1-LIKE; uridine kinase-like 1; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1
Gene Aliases: 1110007H10Rik; F538; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5, (Mouse) Q91YL3
Entrez Gene ID: (Human) 54963, (Mouse) 68556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support