Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EERELSVRAA LASAHQCHPL PRTLSVLKST PQVRGMHTII RDKETSRDEF IFYSKRLMRL LIEHALSFLP FQDCVVQTPQ GQDYAGKCYA GKQITGVSIL
Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UCK1-LIKE; uridine kinase-like 1; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1
Gene Aliases: 1110007H10Rik; F538; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5, (Mouse) Q91YL3
Entrez Gene ID: (Human) 54963, (Mouse) 68556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support