Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is identical in mouse.
Specificity of this antibody: ULK3.
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; Unc-51-like kinase 3
Gene Aliases: 1200015E14Rik; ULK3
UniProt ID: (Human) Q6PHR2, (Mouse) Q3U3Q1
Entrez Gene ID: (Human) 25989, (Mouse) 71742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support