Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Sequence-specific RNA-binding protein which plays an important role in the establishment and maintenance of the early morphology of cortical neurons during embryonic development. Acts as a translation repressor and controls a translationally regulated cell morphology program to ensure proper structuring of the nervous system. Translational control depends on recognition of its binding element within target mRNAs which consists of a mandatory UAG trimer upstream of a U/A-rich motif. Associated with polysomes (PubMed:25737280). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein unkempt homolog; unkempt homolog; Zinc finger CCCH domain-containing protein 5; zinc finger CCCH type containing 5; zinc finger CCCH type domain containing 5; zinc finger CCCH-type containing 5; zinc finger CCCH-type domain containing 5
Gene Aliases: B230379M23Rik; KIAA1753; mKIAA1753; UNK; UNKEMPT; ZC3H5; ZC3HDC5
UniProt ID: (Human) Q9C0B0, (Mouse) Q8BL48
Entrez Gene ID: (Human) 85451, (Mouse) 217331
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support