Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer’s disease, VAMP levels of all isoforms appear to be significantly lowered.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: vamp; VAMP-4; vesicle associated membrane protein 4; Vesicle-associated membrane protein 4
Gene Aliases: D1Ertd147e; VAMP-4; VAMP24; VAMP4
UniProt ID: (Human) O75379, (Mouse) O70480
Entrez Gene ID: (Pig) 100512732, (Human) 8674, (Mouse) 53330, (Rat) 364033
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support