Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EDB; Endobrevin; vamp; VAMP-8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin)
Gene Aliases: AU041171; EDB; endobrevin; VAMP-8; VAMP8
UniProt ID: (Human) Q9BV40, (Mouse) O70404, (Rat) Q9WUF4
Entrez Gene ID: (Human) 8673, (Mouse) 22320, (Rat) 83730
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support