Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALS 8; ALS8VAMP-B/VAMP-C; vamp; VAMP (vesicle-associated membrane protein)-associated protein B and C; Vamp 33b; VAMP B; VAMP B VAMP C; VAMP B/VAMP C; VAMP C; VAMP-associated 33 kDa protein; VAMP-associated protein 33b; VAMP-associated protein B; VAMP-B; VAMP-B/VAMP-C; VAMP-C; VAMPB; VAMPB/VAMPC; VAMPC; VAP 33b; VAP B; VAP B/VAP C; VAP C; VAP-B; VAP-B/VAP-C; VAP-C; VAPB/VAPC; VAPC; vesicle-associated membrane protein, associated protein B and C; Vesicle-associated membrane protein-associated protein B; Vesicle-associated membrane protein-associated protein B/C
Gene Aliases: AI225786; AI840687; AI848259; ALS8; D2Abb2e; R75548; UNQ484/PRO983; VAMP-B; Vamp33b; VAP-B; VAP33b; VAPB
UniProt ID: (Human) O95292, (Rat) Q9Z269
Entrez Gene ID: (Human) 9217, (Rat) 60431, (Mouse) 56491
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support