Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QTDGRNGYT AVIEVTSGGP WGDWAWPEMC PDGFFASGFS LKVEPPQGIP GDDTALNGIR LHCARGNVLG NTHVVESQSG SWGEWSEPLW CRGGAYLVAF SLRVEAPTTL GDNTAANNVR FRCSDGEELQ GPGLSWGDFG DWSDHCPKGA CGLQTKIQGP RGLGDDTALN DARLFCCRS (25-202 aa encoded by BC104195)
VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERGA6350 antibody; MGC125880 antibody; MGC125881 antibody; PRO21055 antibody; vitelline membrane outer layer 1 homolog (chicken) antibody; Vitelline membrane outer layer protein 1 homolog; vitelline membrane outer layer protein 1 homolog antibody; VMO1 antibody
Gene Aliases: ERGA6350; Gm741; PRO21055; RGD1559861; UNQ6350/PRO21055; VMO1
UniProt ID: (Human) Q7Z5L0, (Mouse) Q5SXG7
Entrez Gene ID: (Human) 284013, (Mouse) 327956, (Rat) 360553
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support