Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymal protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; Major epididymis-specific protein E4; MGC57529; Putative protease inhibitor WAP5; WAP domain containing protein HE4-V4; WAP four-disulfide core domain protein 2
Gene Aliases: dJ461P17.6; EDDM4; HE4; WAP5; WFDC2
UniProt ID: (Human) Q14508
Entrez Gene ID: (Human) 10406
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support