Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEKGYSTVTF DGTPSYGHTP SHHAAQFPNH SFKHEDPMGQ QGSLGEQQYS VPPPVYGCHT PTDSCTGSQA LLLRTPYSSD NLYQMTSQLE CMTWNQMNLG ATLKGVAAGS SSSVKWTEGQ SNHSTGYESD NHTTPILCGA QYRMHTHGVF RGIQDVRRVP GVAPTLVRSA SETSEKRPFM CAYPGCNKRY FKLSHLQMHS RKHTGEKPYQ CDFKDCERRF SRSDQLKRHQ RRHTGVKPFQ CKTCQRKFSR SDHLKTHTRT HTGEKPFSCR WPSCQKKFAR SDELVRHHNM HQRNMTKLQL AL
The Wilms Tumor 1 gene encodes one of the WPP domain-interacting proteins (Wilms Tumor 1/AT5G11390, WIT2/AT1G68910) required for RanGAP nuclear envelope association in root tip cells. Ran GTPase plays essential roles in multiple cellular processes, including nucleocytoplasmic transport, spindle formation, and postmitotic nuclear envelope reassembly. The cytoplasmic Ran GTPase activating protein RanGAP is critical to establish a functional RanGTP/RanGDP gradient across the nuclear envelope and is associated with the outer surface of the nuclear envelope in metazoan and higher plant cells. Arabidopsis thaliana RanGAP association with the root tip nuclear envelope requires a family of likely plant-specific nucleoporins combining coiled-coil and transmembrane domains (CC-TMD) and WPP domain-interacting proteins (WIPs). Wilms Tumor 1 and WIT2 have been identified as a second family of CC-TMD proteins, structurally similar, yet clearly distinct from the WIP family, that is required for RanGAP nuclear envelop association in root tip cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Wilms tumor protein; Wilms tumor protein isoform Ex4a(+); WT; WT33
Gene Aliases: AWT1; EWS-WT1; GUD; NPHS4; WAGR; WIT-2; WT1; WT33
UniProt ID: (Human) P19544
Entrez Gene ID: (Human) 7490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support