Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exportin(tRNA); exportin, tRNA (nuclear export receptor for tRNAs); Exportin-T; tRNA exportin
Gene Aliases: 1110004L07Rik; 3110065H13Rik; AI452076; C79645; EXPORTIN-T; XPO3; XPOT
UniProt ID: (Human) O43592
Entrez Gene ID: (Human) 11260, (Mouse) 73192, (Rat) 314879
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support