Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to the XRN1 protein.
Immunogen sequence: SLLREEVRF GGKKTQRVCA PEETTFHLLH LSLMREYIDY EFSVLKEKIT FKYDIERIID DWILMGFLVG NDFIPHLPHL HINHDALPLL YGTYVTILPE LGGYINESGH LNLPRFEKYL VKLSDFDREH FSEVFVDLKW FESKVGNKYL NEAAGVAAEE ARNYKEKKKL KGQENSLCWT ALDKNEGEMI TSKDNLEDET EDDDLFETEF RQYKRTYYMT KMGVDVVSDD FLADQAACYV QAIQWILHYY YHGVQSWSWY YPYHYAPFLS DIHNISTLKI HFELGKPFKP FEQLLAVLPA ASKNLLPACY QHLMTNEDSP IIEYYPPDFK TDLNGKQQEW EAVVLIPFID EKRLLE (222-576 aa encoded by BC078146)
SEC24C is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; 5'-3' exoribonuclease 1; DKFZp434P0721; DKFZp686B22225; DKFZp686F19113; FLJ41903; protein Dhm2; Strand-exchange protein 1 homolog
Gene Aliases: Dhm2; exo; mXrn1; SEP1; XRN1
UniProt ID: (Human) Q8IZH2
Entrez Gene ID: (Human) 54464, (Mouse) 24127
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support