Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Contrin; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; Germ cell-specific Y-box-binding protein; MSY2 homolog; Y box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2
Gene Aliases: CONTRIN; CSDA3; DBPC; MSY2; YBX2
UniProt ID: (Human) Q9Y2T7
Entrez Gene ID: (Human) 51087, (Mouse) 53422
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support