Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PGPGDQEKQQ QASEGRPWSD QAEGPEGEGA MPLFGRTKKR TLGAFSRPPQ RQPVSSRNGL RGVELEASPA QTGNPEETDK LLKRIEVLGF GTVNCGECGL SFSKMTNLLS
May be involved in transcriptional regulation as a repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 133; zinc finger protein 133 (clone pHZ-13); Zinc finger protein 150; zinc finger protein 150 (pHZ-66)
Gene Aliases: pHZ-13; pHZ-66; ZNF133; ZNF150
UniProt ID: (Human) P52736
Entrez Gene ID: (Human) 7692
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support