Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
This antibody recognizes C-terminus (amino acids 434-449 in human) of gamma-tubulin, a 48 kDa structural constituent of cytoskeleton and microtubule organizing center (MTOC). The epitope was located in the amino acid sequence TRPDYI (aa439-444 in human), which is present on human gamma-tubulin 1 but not on human gamma-tubulin 2.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: centrosome marker; Gamma-1-tubulin; Gamma-2-tubulin; Gamma-tubulin complex component 1; GCP-1; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide
Gene Aliases: 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; GCP-1; TUBG; TUBG1; TUBG2; TUBGCP1; Tubgl
UniProt ID: (Human) P23258, (Bovine) Q0VCD2, (Bovine) Q32KM1, (Human) Q9NRH3, (Mouse) P83887, (Rat) P83888, (Mouse) Q8VCK3
Entrez Gene ID: (Human) 7283, (Chicken) 420025, (Bovine) 541101, (Bovine) 540216, (Human) 27175, (Mouse) 103733, (Rat) 252921, (Rat) 680991, (Mouse) 103768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support