Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
This antibody can label the centrosome structure.
Immunogen sequence: GSYLLERLN DRYPKKLVQT YSVFPNQDEM SDVVVQPYNS LLTLKRLTQN ADCVVVLDNT ALNRIATDRL HIQNPSFSQI NQLVSTIMSA STTTLRYPGY MNNDLIGLIA SLIPTPRLHF LMTGYTPLTT DQSVASVRKT TVLDVMRRLL QPKNVMVSTG RDRQTNHCYI AILNIIQGEV DPTQVHKSLQ RIRERKLANF IPWGPASIQV ALSRKSPYLP SAHRVSGLMM ANHTSISSLF ERTCRQYDKL RKREAFLEQF RKEDMFKDNF DEMDTSREIV QQLIDEYHAA TRPDYISWGT QEQ (150-451 aa encoded by BC000619)
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosome marker; gamma tubulin; Gamma-1-tubulin; Gamma-2-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide
Gene Aliases: 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; GCP-1; TUBG; TUBG1; TUBG2; TUBGCP1; Tubgl
UniProt ID: (Human) P23258, (Dog) Q9GKK5, (Human) Q9NRH3, (Mouse) P83887, (Mouse) Q8VCK3
Entrez Gene ID: (Human) 7283, (Dog) 403695, (Human) 27175, (Mouse) 103733, (Mouse) 103768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support