Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Laminin antibodies are widely used to label blood vessels and basement membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystrophia muscularis; Laminin A chain; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1 (formerly LAMB2); Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma
Gene Aliases: 5830440B04; AA408497; C77966; C80098; C81607; CLM; D130003D08Rik; dy; LAMA; Lama-1; LAMA1; LAMA2; Lamb-1; LAMB1; Lamb1-1; LAMB2; LAMC1; LAMM; LIS5; mer; merosin; mKIAA4087; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047, (Mouse) P19137, (Mouse) Q60675, (Mouse) P02469
Entrez Gene ID: (Human) 284217, (Human) 3908, (Human) 3912, (Human) 3915, (Mouse) 16772, (Mouse) 16773, (Mouse) 16777, (Mouse) 226519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support