Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 16 amino acids near the center of human CENPO.
PEP-0940 can be used as a blocking peptide with polyclonal antibody PA5-20826.
Accurate chromosome segregation during mitosis requires a kinetochore to assemble correctly at the centromere of each chromatid and form a dynamic interface with the microtubules of the mitotic spindle. The kinetochore assembly includes the multisubunit CENP-H/I complex which can be divided into three functional classes based on phenotype analysis. One of these classes is referred to as the CENP-O class of proteins of which CENPO is the exemplar. These proteins form a stable complex with each other and are required for proper kinetochore function and for recovery from mitotic spindle damage. CENPO has also been shown to be an auto-antigen in a small population of anti-centromere antibodies- (ACA-) positive patients with sclerodoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-O; Centromere protein O; centromeric protein O; Interphase centromere complex protein 36
Gene Aliases: CENP-O; CENPO; ICEN-36; ICEN36; MCM21R
UniProt ID: (Human) Q9BU64
Entrez Gene ID: (Human) 79172
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support